Learn More
Creative Biomart Recombinant HCV NS3 Antigen, His-tagged
Supplier:  Creative Biomart HCV198
Encompass_Preferred
The E. coli derived recombinant protein contains a total of 268 amino acids having a MW of 27.9 kDa. The protein contains the HCV NS3 immunodominant regions, amino acids 1225-1456. The HCV NS3 protein
Catalog No. 50-208-4576