Learn More
Creative Biomart Recombinant HCV NS4 a+b Antigen, b-galactosidase-tagged
Supplier:  Creative Biomart HCV202
Encompass_Preferred
The E. coli derived 19 kDa recombinant protein contains the HCV NS4 immunodominant regions, amino acids 1658-1863. The protein is fused with b-galactosidase (114 kDa) at N-terminus, pI 5.45.
Catalog No. 50-208-4580