Learn More
Creative Biomart Recombinant HCV NS5B Antigen, His-tagged
Supplier:  Creative Biomart HCV230
Encompass_Preferred
The E. coli derived recombinant protein contains the full length of the NS5B immunodominant regions Genotype 1B and fused to a 6 His tag at C-Terminus.
Catalog No. 50-208-4608