Learn More
Andwin Scientific HD 20L CARBOY W/SPIGOT PP EA
SDP
Catalog No. 501305871
Encompass_Preferred
The 20-liter carboy features a leakproof spigot, an octagonal design that increases rigidity when full, and graduations with 1 L increments that allow accurate delivery and measurement of liquids. This product includes a polypropylene closure that lessens evaporation. It is also autoclavable up to 275F.
Catalog No. 50-130-5871 Supplier Andwin Scientific Supplier No. 6250725A
The 20-liter carboy features a leakproof spigot, an octagonal design that increases rigidity when full, and graduations with 1 L increments that allow accurate delivery and measurement of liquids. This product includes a polypropylene closure that lessens evaporation. It is also autoclavable up to 275F.