Learn More
Cole-Parmer Cole-Parmer Economical heavy duty carboy with spigot, 20 liter
Catalog No. NC9368344 Shop All Cole-Parmer Products
Encompass_Preferred
Heavy-duty graduated polypropylene (PP) carboys feature octagonal shape for increased rigidity when full. The neck is offset for easier filling and pouring. Molded handle is sealed at the ends to prevent contamination. Durable, leakproof spigot is supplied with a tube fitting to accept 0.31" (8-mm) ID tubing. Carboys include a PP screw closure. Autoclavable up to 275°F.
Catalog No. NC9368344 Supplier Cole-Parmer Supplier No. 6250725
Heavy-duty graduated polypropylene (PP) carboys feature octagonal shape for increased rigidity when full. The neck is offset for easier filling and pouring. Molded handle is sealed at the ends to prevent contamination. Durable, leakproof spigot is supplied with a tube fitting to accept 0.31" (8-mm) ID tubing. Carboys include a PP screw closure. Autoclavable up to 275°F.