Learn More
MSC Baldwin Filters Air Breather Element, Round, Radial Seal, 1 ea
Catalog No. 5000508035
Encompass
This heavy-duty air breather element is designed to prevent contaminants and water from entering hydraulic or fluid systems during reservoir fluctuations. It also helps prevent ambient humidity from condensing within tanks, ensuring cleaner fluid and protected equipment operation.
  • Designed for radial seal applications
  • Round filter shape
  • Constructed from cellulose and paper media
  • Includes a 1 3/8 in. gasket
  • Compatible with diverse heavy equipment and truck applications
  • ISO 9001:2015 certified
Catalog No. 50-005-08035 Supplier MSC Supplier No. 55873723
This heavy-duty air breather element is designed to prevent contaminants and water from entering hydraulic or fluid systems during reservoir fluctuations. It also helps prevent ambient humidity from condensing within tanks, ensuring cleaner fluid and protected equipment operation.
  • Designed for radial seal applications
  • Round filter shape
  • Constructed from cellulose and paper media
  • Includes a 1 3/8 in. gasket
  • Compatible with diverse heavy equipment and truck applications
  • ISO 9001:2015 certified