Learn More
MSC Baldwin Air Element, Axial Seal, 62.7 mm Length, 1 ea
Catalog No. 5000509779
Encompass
These axial seal air filter elements maintain optimal equipment performance by effectively preventing airborne contaminants from entering the combustion chamber. Designed for durability in demanding environments, the filters feature securely affixed metal end caps and a raised gasket to provide a reliable seal and prevent bypass leakage.
  • Prevents airborne contaminants from reaching the combustion chamber
  • Features securely affixed metal end caps
  • Incorporates a raised gasket to ensure a proper seal
  • Compatible with Benford equipment and Lister engines
  • Dimensions include an outside diameter of 142.1 mm (5 19/32 in.), inside diameter of 101.6 mm (4 in.), and height of 62.7 mm (2 15/32 in.)
Catalog No. 50-005-09779 Supplier MSC Supplier No. 55989883
These axial seal air filter elements maintain optimal equipment performance by effectively preventing airborne contaminants from entering the combustion chamber. Designed for durability in demanding environments, the filters feature securely affixed metal end caps and a raised gasket to provide a reliable seal and prevent bypass leakage.
  • Prevents airborne contaminants from reaching the combustion chamber
  • Features securely affixed metal end caps
  • Incorporates a raised gasket to ensure a proper seal
  • Compatible with Benford equipment and Lister engines
  • Dimensions include an outside diameter of 142.1 mm (5 19/32 in.), inside diameter of 101.6 mm (4 in.), and height of 62.7 mm (2 15/32 in.)