Learn More
MSC Baldwin Air Element, Heavy Duty, Axial Seal, 1 ea
Catalog No. 5000509783
Encompass
The PA2938 Axial Seal Heavy Duty Air Filter Element is designed to maintain optimal engine performance by preventing airborne contaminants from entering the combustion chamber. This filter is suitable for various industrial applications and features a robust construction to ensure equipment protection in demanding environments.
  • Designed for industrial applications
  • Features axial seal technology
  • Includes an attached gasket for reliable installation
  • Outside diameter: 204.0 mm (8 1/32 in.)
  • Inside diameter: 154.0 mm (6 1/16 in.)
  • Length: 304.8 mm (12 in.)
Catalog No. 50-005-09783 Supplier MSC Supplier No. 55990626
The PA2938 Axial Seal Heavy Duty Air Filter Element is designed to maintain optimal engine performance by preventing airborne contaminants from entering the combustion chamber. This filter is suitable for various industrial applications and features a robust construction to ensure equipment protection in demanding environments.
  • Designed for industrial applications
  • Features axial seal technology
  • Includes an attached gasket for reliable installation
  • Outside diameter: 204.0 mm (8 1/32 in.)
  • Inside diameter: 154.0 mm (6 1/16 in.)
  • Length: 304.8 mm (12 in.)