Learn More
MSC Baldwin Conical-Shaped Air Element, 495.3 mm Height, Attached Gasket, 1 ea
Catalog No. 5000509675
Encompass
This conical-shaped air element is designed to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. It is compatible with ERF and Foden trucks and includes an attached gasket for reliable sealing.
  • Conical design for precise fitment
  • Features securely affixed metal end caps
  • Integrated axial seal for improved filtration performance
  • Includes gasket G103-N
  • Engineered for heavy-duty applications
  • Compatible with replacement parts ERF 990199 and Foden Y05590000
Catalog No. 50-005-09675 Supplier MSC Supplier No. 55984512
This conical-shaped air element is designed to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. It is compatible with ERF and Foden trucks and includes an attached gasket for reliable sealing.
  • Conical design for precise fitment
  • Features securely affixed metal end caps
  • Integrated axial seal for improved filtration performance
  • Includes gasket G103-N
  • Engineered for heavy-duty applications
  • Compatible with replacement parts ERF 990199 and Foden Y05590000