Learn More
MSC Baldwin Filters Air Element, Round, 17 1/4 in Height, 1 ea
Catalog No. 5000508683
Encompass
This heavy-duty round engine air filter element is designed to provide effective filtration for various industrial machinery applications. It ensures optimal engine performance by trapping particulate matter before it enters the air intake system.
  • Round air filter element design
  • Includes attached gasket for secure seating
  • Features integrated lift tab for ease of maintenance
  • Compatible with equipment from Dresser, Drott, Elgin, International, and Wayne
  • Measures 139.7 mm (5 1/2 in.) in outside diameter and 79.4 mm (3 1/8 in.) in inside diameter
  • Total height of 438.2 mm (17 1/4 in.)
Catalog No. 50-005-08683 Supplier MSC Supplier No. 55987119
This heavy-duty round engine air filter element is designed to provide effective filtration for various industrial machinery applications. It ensures optimal engine performance by trapping particulate matter before it enters the air intake system.
  • Round air filter element design
  • Includes attached gasket for secure seating
  • Features integrated lift tab for ease of maintenance
  • Compatible with equipment from Dresser, Drott, Elgin, International, and Wayne
  • Measures 139.7 mm (5 1/2 in.) in outside diameter and 79.4 mm (3 1/8 in.) in inside diameter
  • Total height of 438.2 mm (17 1/4 in.)