Learn More
MSC Baldwin Filters Replacement Ecolite Air Element, 168.3 mm OD, 50.8 mm ID, 262.7 mm Length, 1 ea
Catalog No. 5000508633
Encompass
The PA2803 HD Air Element is a heavy-duty replacement filter designed for use in disposable housings, serving as an efficient alternative for specific refrigeration equipment.
  • Designed as a replacement for Ecolite air elements
  • Suitable for use in Carrier-Transicold refrigeration units
  • Outside diameter of 168.3 mm
  • Inside diameter of 50.8 mm
  • Length of 262.7 mm
  • Compatible with Carrier 30-01077-01
Catalog No. 50-005-08633 Supplier MSC Supplier No. 55984827
The PA2803 HD Air Element is a heavy-duty replacement filter designed for use in disposable housings, serving as an efficient alternative for specific refrigeration equipment.
  • Designed as a replacement for Ecolite air elements
  • Suitable for use in Carrier-Transicold refrigeration units
  • Outside diameter of 168.3 mm
  • Inside diameter of 50.8 mm
  • Length of 262.7 mm
  • Compatible with Carrier 30-01077-01