Learn More
MSC Baldwin Filters Air Element, Dual-Stage, Round, Cellulose Paper, 1 ea.
Catalog No. 5000509905
Encompass
This heavy-duty automotive air filter is a dual-stage element designed to protect the combustion chamber by preventing airborne contaminants from entering the engine. Featuring a robust, round axial seal design, it incorporates securely affixed metal end caps with a raised gasket to ensure a proper seal and eliminate leaks.
  • Dual-stage design includes a protective blanket and screen
  • Round axial seal construction ensures high performance and prevents bypass
  • Constructed from high-quality cellulose and paper media
  • Manufactured under ISO 9001:2015 standards
  • Engineered for specific vehicle applications, including select trucks and equipment
Catalog No. 50-005-09905 Supplier MSC Supplier No. 55987093
This heavy-duty automotive air filter is a dual-stage element designed to protect the combustion chamber by preventing airborne contaminants from entering the engine. Featuring a robust, round axial seal design, it incorporates securely affixed metal end caps with a raised gasket to ensure a proper seal and eliminate leaks.
  • Dual-stage design includes a protective blanket and screen
  • Round axial seal construction ensures high performance and prevents bypass
  • Constructed from high-quality cellulose and paper media
  • Manufactured under ISO 9001:2015 standards
  • Engineered for specific vehicle applications, including select trucks and equipment