Learn More
MSC Baldwin Axial Seal Air Filter Element, 301.6 mm OD, 673.1 mm Height, 1 ea
Catalog No. 5000509942
Encompass
This axial seal air filter element is designed for heavy-duty filtration applications, ensuring high performance and efficient protection of equipment air intake systems. The filter features a robust construction with metal end caps and a raised, attached gasket to provide a reliable seal.
  • Designed with an axial seal for secure fitment
  • Equipped with durable metal end caps
  • Features a raised, attached gasket
  • Measures 301.6 mm (11 7/8 in.) in outside diameter and 673.1 mm (26 1/2 in.) in overall height
Catalog No. 50-005-09942 Supplier MSC Supplier No. 55990618
This axial seal air filter element is designed for heavy-duty filtration applications, ensuring high performance and efficient protection of equipment air intake systems. The filter features a robust construction with metal end caps and a raised, attached gasket to provide a reliable seal.
  • Designed with an axial seal for secure fitment
  • Equipped with durable metal end caps
  • Features a raised, attached gasket
  • Measures 301.6 mm (11 7/8 in.) in outside diameter and 673.1 mm (26 1/2 in.) in overall height