Learn More
MSC HD ALKALINE CLEANER
Catalog No. 502818985
Encompass
ZEP Tile & Grout Cleaner is an alkaline cleaner specifically formulated for effective cleaning in bath, restroom, and shower environments.
  • Liquid formulation
  • Pleasant scent
  • Designed for bathroom cleaning applications
  • High alkaline pH (12.5-13.5)
  • Packaged as 12 one-quart bottles per case
Catalog No. 50-281-8985 Supplier MSC Supplier No. 17595380
ZEP Tile & Grout Cleaner is an alkaline cleaner specifically formulated for effective cleaning in bath, restroom, and shower environments.
  • Liquid formulation
  • Pleasant scent
  • Designed for bathroom cleaning applications
  • High alkaline pH (12.5-13.5)
  • Packaged as 12 one-quart bottles per case