Learn More
MSC Baldwin Filters Cabin Air Filter, Rectangular, 37 in. Length, 1 ea.
Catalog No. 5000509634
Encompass
This cabin air filter element is designed to remove contaminants and pollutants from the air before they enter the vehicle cabin, helping to maintain a safe and comfortable environment for operators. Its space-saving, rectangular panel design is suitable for use in a variety of heavy-duty trucks and passenger vehicles.
  • Designed to help prevent pollutants from entering the cabin area
  • Rectangular panel design for space-efficient installation
  • Compatible with John Deere and White Equipment vehicles
  • Replaces John Deere AH80212
Catalog No. 50-005-09634 Supplier MSC Supplier No. 55986657
This cabin air filter element is designed to remove contaminants and pollutants from the air before they enter the vehicle cabin, helping to maintain a safe and comfortable environment for operators. Its space-saving, rectangular panel design is suitable for use in a variety of heavy-duty trucks and passenger vehicles.
  • Designed to help prevent pollutants from entering the cabin area
  • Rectangular panel design for space-efficient installation
  • Compatible with John Deere and White Equipment vehicles
  • Replaces John Deere AH80212