Learn More
MSC Smith Performance Sprayers R300 Sprayer, 3 gal., Polyethylene, 1 ea
Catalog No. 502867101
Encompass
The Smith Performance Sprayers R300 is a professional-grade 3-gallon compression sprayer designed for robust chemical applications, featuring durable components for demanding agricultural and industrial use.
  • Durable 21 in stainless steel wand with poly lining and Viton seals for maximum chemical resistance.
  • Highly efficient pump for fewer pumps per gallon.
  • In-line, easy-to-access filter reduces clogging.
  • Pump seals can be changed quickly without tools for faster repair.
  • Includes 5 versatile nozzles: brass and poly adjustable, wide and narrow flat fans, and foaming.
  • Polyethylene tank material for chemical compatibility.
  • Deluxe shoulder harness with nozzle storage and back support cushion.
  • Chemical-resistant 50 in hose for durability.
  • Can be regulated to 25 PSI pressure.
Catalog No. 50-286-7101 Supplier MSC Supplier No. 30302368
The Smith Performance Sprayers R300 is a professional-grade 3-gallon compression sprayer designed for robust chemical applications, featuring durable components for demanding agricultural and industrial use.
  • Durable 21 in stainless steel wand with poly lining and Viton seals for maximum chemical resistance.
  • Highly efficient pump for fewer pumps per gallon.
  • In-line, easy-to-access filter reduces clogging.
  • Pump seals can be changed quickly without tools for faster repair.
  • Includes 5 versatile nozzles: brass and poly adjustable, wide and narrow flat fans, and foaming.
  • Polyethylene tank material for chemical compatibility.
  • Deluxe shoulder harness with nozzle storage and back support cushion.
  • Chemical-resistant 50 in hose for durability.
  • Can be regulated to 25 PSI pressure.