Learn More
MSC Nu-Calgon Drain Cleaner, Liquid, 1 quart, Unscented, Alkaline
Catalog No. 502836661
Encompass
Nu-Calgon Drain Solve is an alkaline-based drain opener designed to remove grease, hair, vegetable, and other organic matter from drains. It is non-fuming and quickly cleans and clears drains without fear of noxious fumes or reactive chemicals. This product is NSF registered and does not contain sulfuric acid.
  • Perfect for clogged A/C condensate lines, kitchen, bathtub, and lavatory drains
  • Non-fuming
  • No violent eruptions
  • Pours through standing water
  • Cuts through grease and organic matter
  • Safe on septic systems
  • Alkaline-based drain opener
  • Cleans and clears drains quickly
  • Does not contain sulfuric acid
  • NSF registered
  • Unscented
Catalog No. 50-283-6661 Supplier MSC Supplier No. 77410496
Nu-Calgon Drain Solve is an alkaline-based drain opener designed to remove grease, hair, vegetable, and other organic matter from drains. It is non-fuming and quickly cleans and clears drains without fear of noxious fumes or reactive chemicals. This product is NSF registered and does not contain sulfuric acid.
  • Perfect for clogged A/C condensate lines, kitchen, bathtub, and lavatory drains
  • Non-fuming
  • No violent eruptions
  • Pours through standing water
  • Cuts through grease and organic matter
  • Safe on septic systems
  • Alkaline-based drain opener
  • Cleans and clears drains quickly
  • Does not contain sulfuric acid
  • NSF registered
  • Unscented