Learn More
Grainger Checkers Safety™ Warning Whip Flag, Nylon Mesh, Yellow, 12 in., 1 ea.
Catalog No. 502788475
Encompass_Preferred
This heavy-duty, fluorescent Day Bright™ nylon mesh flag is designed for use with warning whip systems, offering high visibility for enhanced safety. The flag features a white reflective X for increased awareness in low-light conditions and includes necessary mounting clips for easy installation.
  • Fluorescent nylon mesh material for high-visibility applications
  • Equipped with a white reflective X for improved visibility
  • Includes mounting clips for secure attachment
  • Suitable for use with various warning whip models
Catalog No. 50-278-8475 Supplier Grainger Supplier No. 16D833
This heavy-duty, fluorescent Day Bright™ nylon mesh flag is designed for use with warning whip systems, offering high visibility for enhanced safety. The flag features a white reflective X for increased awareness in low-light conditions and includes necessary mounting clips for easy installation.
  • Fluorescent nylon mesh material for high-visibility applications
  • Equipped with a white reflective X for improved visibility
  • Includes mounting clips for secure attachment
  • Suitable for use with various warning whip models