Learn More
MSC Baldwin Filters Foam Blanket, Rectangular, 1531.9 mm x 241.3 mm x 12.7 mm, 1 ea
Catalog No. 5000509909
Encompass
This foam blanket serves as a high-density prefilter component designed for integration with compatible air filter systems. It provides an essential layer of protection to help maintain air filtration performance.
  • Designed for use with PA2653 air filters
  • Constructed from durable foam material
  • Rectangular shape
  • Dimensions measure 1,531.9 mm x 241.3 mm x 12.7 mm (60 5/16 in. x 9 1/2 in. x 1/2 in.)
Catalog No. 50-005-09909 Supplier MSC Supplier No. 55990311
This foam blanket serves as a high-density prefilter component designed for integration with compatible air filter systems. It provides an essential layer of protection to help maintain air filtration performance.
  • Designed for use with PA2653 air filters
  • Constructed from durable foam material
  • Rectangular shape
  • Dimensions measure 1,531.9 mm x 241.3 mm x 12.7 mm (60 5/16 in. x 9 1/2 in. x 1/2 in.)