Learn More
MSC  Baldwin Filters, HDFUEL ELEMENT
Catalog No. 5000508249
Encompass
This heavy duty fuel element is designed for use with DAHL 2 series fuel filter/water separator units. It provides effective filtration for fuel systems, ensuring reliable engine performance.
  • Designed for DAHL 200 series fuel filter/water separator units
  • Nominal micron rating of 4
  • Absolute micron rating of 25
  • Includes attached gasket and O-ring
  • Dimensions of 117.5 mm (4 5/8 in.) length and 154.0 mm (6 1/16 in.) outside diameter
Catalog No. 50-005-08249 Supplier MSC Supplier No. 55862957
This heavy duty fuel element is designed for use with DAHL 2 series fuel filter/water separator units. It provides effective filtration for fuel systems, ensuring reliable engine performance.
  • Designed for DAHL 200 series fuel filter/water separator units
  • Nominal micron rating of 4
  • Absolute micron rating of 25
  • Includes attached gasket and O-ring
  • Dimensions of 117.5 mm (4 5/8 in.) length and 154.0 mm (6 1/16 in.) outside diameter