Learn More
MSC Baldwin Filters Fuel Filter Element, Cellulose, 9.8 Micron, 1 ea
Catalog No. 5000508404
Encompass
The PF926 is a high-performance cartridge-style filter element designed for diesel fuel and hydraulic systems. Constructed from cellulose, this filter element is engineered to provide efficient filtration for a variety of heavy-duty applications, ensuring optimal fluid cleanliness and system protection.
  • Designed for use in diesel fuel and hydraulic systems
  • Features a cartridge filter design with an attached gasket
  • Meets ISO 9001:2015 quality standards
  • Compatible with specific diesel engines found in trucks and buses
  • Measures 2 9/16 in. outside diameter and 9 3/4 in. height
  • Inside diameter measures 1 1/16 in. (27.0 mm)
Catalog No. 50-005-08404 Supplier MSC Supplier No. 55866024
The PF926 is a high-performance cartridge-style filter element designed for diesel fuel and hydraulic systems. Constructed from cellulose, this filter element is engineered to provide efficient filtration for a variety of heavy-duty applications, ensuring optimal fluid cleanliness and system protection.
  • Designed for use in diesel fuel and hydraulic systems
  • Features a cartridge filter design with an attached gasket
  • Meets ISO 9001:2015 quality standards
  • Compatible with specific diesel engines found in trucks and buses
  • Measures 2 9/16 in. outside diameter and 9 3/4 in. height
  • Inside diameter measures 1 1/16 in. (27.0 mm)