Learn More
MSC Baldwin Fuel Spin-on, 4 Absolute Micron, M16 x 1.5 Thread, 1 ea
Catalog No. 5000508937
Encompass
The BF7919 fuel spin-on filter is engineered to maintain engine efficiency by effectively removing fuel contaminants. Designed for heavy-duty applications, this spin-on unit ensures fuel cleanliness, which is essential for modern injection systems.
  • Absolute filtration rating of 4 microns
  • Thread size: M16 x 1.5
  • Outside diameter: 93.7 mm (3 11/16 in.)
  • Length: 133.4 mm (5 1/4 in.)
  • Gasket included (G381-A)
  • Compatible with John Deere equipment (replaces AT279824)
Catalog No. 50-005-08937 Supplier MSC Supplier No. 55863732
The BF7919 fuel spin-on filter is engineered to maintain engine efficiency by effectively removing fuel contaminants. Designed for heavy-duty applications, this spin-on unit ensures fuel cleanliness, which is essential for modern injection systems.
  • Absolute filtration rating of 4 microns
  • Thread size: M16 x 1.5
  • Outside diameter: 93.7 mm (3 11/16 in.)
  • Length: 133.4 mm (5 1/4 in.)
  • Gasket included (G381-A)
  • Compatible with John Deere equipment (replaces AT279824)