Learn More
MSC Baldwin Fuel/Water Separator Spin-on, Cellulose, 4 Micron, 1 ea
Catalog No. 5000508361
Encompass
This heavy-duty spin-on fuel/water separator is designed to protect modern fuel injection systems by effectively removing particulate and water contamination, ensuring engines maintain maximum efficiency. Constructed with cellulose media, it provides robust filtration for demanding environments.
  • Features a drain for easy water removal
  • Includes an integral G401 gasket and post seal
  • Engineered for use in diesel and gasoline engine applications
  • Compatible with Caterpillar engines
  • Meets ISO 9001:2015 quality standards
  • Designed for vehicle-specific fitment in trucks and buses
Catalog No. 50-005-08361 Supplier MSC Supplier No. 55862510
This heavy-duty spin-on fuel/water separator is designed to protect modern fuel injection systems by effectively removing particulate and water contamination, ensuring engines maintain maximum efficiency. Constructed with cellulose media, it provides robust filtration for demanding environments.
  • Features a drain for easy water removal
  • Includes an integral G401 gasket and post seal
  • Engineered for use in diesel and gasoline engine applications
  • Compatible with Caterpillar engines
  • Meets ISO 9001:2015 quality standards
  • Designed for vehicle-specific fitment in trucks and buses