Learn More
MSC Baldwin Filters Engine Air Filter, Round, Axial Seal, Cellulose, 1 ea
Catalog No. 5000508613
Encompass
This automotive engine inner air filter is designed for use in farm equipment, including Massey Ferguson wheel tractors. Constructed from cellulose and paper media, the round axial seal element prevents airborne contaminants from entering the engine combustion chamber to maintain optimal performance.
  • Features securely affixed metal end caps with a raised gasket for a proper seal and leak prevention
  • Designed for use as an inner air filter element
  • Compatible with models PA2604 and PA2611
  • Includes an attached gasket
  • Meets ISO 9001:2015 standards
  • Height: 14 3/8 in. (36.53 cm)
  • Width: 6 7/8 in. (17.48 cm)
Catalog No. 50-005-08613 Supplier MSC Supplier No. 55986384
This automotive engine inner air filter is designed for use in farm equipment, including Massey Ferguson wheel tractors. Constructed from cellulose and paper media, the round axial seal element prevents airborne contaminants from entering the engine combustion chamber to maintain optimal performance.
  • Features securely affixed metal end caps with a raised gasket for a proper seal and leak prevention
  • Designed for use as an inner air filter element
  • Compatible with models PA2604 and PA2611
  • Includes an attached gasket
  • Meets ISO 9001:2015 standards
  • Height: 14 3/8 in. (36.53 cm)
  • Width: 6 7/8 in. (17.48 cm)