Learn More
MSC Sweepex Magnet Bar, 60 in, Metal, Silver/Red
Catalog No. 502836715
Encompass
This magnet bar is designed for collecting magnetic debris. It features a durable metal construction and an adjustable chain and hook system for easy attachment to forklift brooms, ensuring efficient cleanup in various industrial settings.
  • Durable metal construction
  • Adjustable chain and hook mounting system
  • Attaches in front of Mega and Pro Series forklift brooms
  • 60 in overall sweeper width
  • 4 in overall depth
  • 2 in overall height
Catalog No. 50-283-6715 Supplier MSC Supplier No. 46051934
This magnet bar is designed for collecting magnetic debris. It features a durable metal construction and an adjustable chain and hook system for easy attachment to forklift brooms, ensuring efficient cleanup in various industrial settings.
  • Durable metal construction
  • Adjustable chain and hook mounting system
  • Attaches in front of Mega and Pro Series forklift brooms
  • 60 in overall sweeper width
  • 4 in overall depth
  • 2 in overall height