Learn More
Med Vet International Jorvet Equine Padded Recovery Hood, with Protective Foam and Adjustable Strap, J0473
Catalog No. 5000495342
Encompass_Preferred
This protective hood is a valuable tool in the prevention of head trauma such as facial nerve paralysis, to any horse undergoing anesthesia. The thick yellow closed-cell foam padding has openings for eyes, ears, and muzzles. The area around the eyes is double padded.
Catalog No. 50-004-95342 Supplier Med Vet International Supplier No. JOR473
This protective hood is a valuable tool in the prevention of head trauma such as facial nerve paralysis, to any horse undergoing anesthesia. The thick yellow closed-cell foam padding has openings for eyes, ears, and muzzles. The area around the eyes is double padded.