Learn More
MSC Baldwin Outer Air Element, Axial Seal, 12 15/16 in. Length, 1 ea.
Catalog No. 5000509891
Encompass
The PA2651 heavy-duty outer air element is designed for use as a reliable filtration component in demanding operational environments, specifically compatible with Komatsu equipment. It serves as a critical barrier to maintain engine health by capturing particulate matter from the air intake system.
  • Fits Komatsu equipment
  • Includes attached gasket for secure installation
  • Axial seal design ensures optimal performance
  • Compatible with inner air element PA2652
  • Dimensions: 209.6 mm (8 1/4 in.) outside diameter, 328.6 mm (12 15/16 in.) length
Catalog No. 50-005-09891 Supplier MSC Supplier No. 55986285
The PA2651 heavy-duty outer air element is designed for use as a reliable filtration component in demanding operational environments, specifically compatible with Komatsu equipment. It serves as a critical barrier to maintain engine health by capturing particulate matter from the air intake system.
  • Fits Komatsu equipment
  • Includes attached gasket for secure installation
  • Axial seal design ensures optimal performance
  • Compatible with inner air element PA2652
  • Dimensions: 209.6 mm (8 1/4 in.) outside diameter, 328.6 mm (12 15/16 in.) length