Learn More
MSC Baldwin Filters Engine Air Filter, Outer Element, 13 13/16 in. Diameter, 22 1/2 in. Height, 1 ea.
Catalog No. 5000508590
Encompass
This heavy-duty outer air filter element is designed as an engine air filter to protect combustion chambers by preventing airborne contaminants from entering the system. The unit features an axial seal design, incorporating securely affixed metal end caps and a raised gasket to ensure a robust, leak-proof installation.
  • Outer air filter element design
  • Includes two attached gaskets for secure sealing
  • Engineered for use in compatible heavy equipment including Allis Chalmers, Euclid, Fiat-Allis, Terex, and Versatile units
  • Outside diameter of 13 13/16 in. (350.8 mm)
  • Overall height of 22 1/2 in. (571.5 mm)
Catalog No. 50-005-08590 Supplier MSC Supplier No. 55985048
This heavy-duty outer air filter element is designed as an engine air filter to protect combustion chambers by preventing airborne contaminants from entering the system. The unit features an axial seal design, incorporating securely affixed metal end caps and a raised gasket to ensure a robust, leak-proof installation.
  • Outer air filter element design
  • Includes two attached gaskets for secure sealing
  • Engineered for use in compatible heavy equipment including Allis Chalmers, Euclid, Fiat-Allis, Terex, and Versatile units
  • Outside diameter of 13 13/16 in. (350.8 mm)
  • Overall height of 22 1/2 in. (571.5 mm)