Learn More
MSC ZEP Oven Cleaner, Liquid, Lemon Scent, Drum, 55 gal
Catalog No. 502819264
Encompass
This ZEP On & Off Liquid Oven & Grill Cleaner is a heavy-duty, ready-to-use liquid cleaner designed for effective removal of grease and baked-on food from various surfaces. It is formulated with a lemon scent and is ideal for commercial and industrial kitchen applications, featuring a pH level between 13 and 14.
  • Ready-to-use, water-based formula
  • Lemon fragrance
  • Nonflammable and phosphate-free
  • Butyl-free
  • Effective temperature of 200 degrees F
  • NSF A8 rated
  • For use on drip pans, grills, hoods, ovens, and stoves
Catalog No. 50-281-9264 Supplier MSC Supplier No. 17596016
This ZEP On & Off Liquid Oven & Grill Cleaner is a heavy-duty, ready-to-use liquid cleaner designed for effective removal of grease and baked-on food from various surfaces. It is formulated with a lemon scent and is ideal for commercial and industrial kitchen applications, featuring a pH level between 13 and 14.
  • Ready-to-use, water-based formula
  • Lemon fragrance
  • Nonflammable and phosphate-free
  • Butyl-free
  • Effective temperature of 200 degrees F
  • NSF A8 rated
  • For use on drip pans, grills, hoods, ovens, and stoves