Learn More
Research Products International Corp High Density Polyethylene MiniVial, 7ml Capacity, 17mm Cap Size, Bulk Pack, 1000 per Case
SDP
Catalog No. 50550150
Encompass
Rigid linear polyethylene vial with high resistance to solvent permeation. Supplied with an unlined polyethylene vial with screw cap. Bulk pack contains 1,000 vials and caps packaged separately in clear plastic bags.
Catalog No. 50-550-150 Supplier Research Products International Corp Supplier No. 125500
Rigid linear polyethylene vial with high resistance to solvent permeation. Supplied with an unlined polyethylene vial with screw cap. Bulk pack contains 1,000 vials and caps packaged separately in clear plastic bags.