Learn More
Azzota Corp Heavy-Duty Portable Refractometer, Sturdy Design, Made Of Aluminum Brix Range: 0-10%, Brix Min.Div.: 1% Brix Accuracy: +/-1.0%, With Atc
Catalog No. 501612923
Encompass_Preferred
Heavy-Duty Portable Refractometer, Sturdy design, made of Aluminum Brix Range: 0-10%, Brix Min.Div.: 1% Brix Accuracy: +/-1.0%, With ATC
Catalog No. 50-161-2923 Supplier Azzota Corp Supplier No. RHB10T
Heavy-Duty Portable Refractometer, Sturdy design, made of Aluminum Brix Range: 0-10%, Brix Min.Div.: 1% Brix Accuracy: +/-1.0%, With ATC