Learn More
Azzota Corp Heavy-Duty Portable Wine Tester Refractometer. With Atc. Oe Range 30-140, Mas Sacch Range 0-32%, Min. Div. Oe 1, Mas Sacch 0.1%
Catalog No. 501612925
Encompass_Preferred
Heavy-Duty Portable Wine Tester Refractometer. With ATC. Oe range 30-140, Mas Sacch range 0-32%, Min. Div. Oe 1, Mas Sacch 0.1%
Catalog No. 50-161-2925 Supplier Azzota Corp Supplier No. RHW3
Heavy-Duty Portable Wine Tester Refractometer. With ATC. Oe range 30-140, Mas Sacch range 0-32%, Min. Div. Oe 1, Mas Sacch 0.1%