Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP All-Purpose Cleaner, Granules, Powder, Pine Scent, 40 lb

Catalog No. 502869001
Encompass

The All-Purpose Cleaner is designed to effectively remove tough, embedded soils from concrete floors. It comes in a granular or powder form and dissolves quickly for efficient cleaning.

  • Form: Granules, powder
  • Container type: Pail
  • Container size: 40 lb
  • Scent: Pine
  • Application: Dissolves quickly and removes tough, embedded soils from concrete floors

Catalog No. 50-286-9001 Supplier MSC Supplier No. 13836259
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The All-Purpose Cleaner is designed to effectively remove tough, embedded soils from concrete floors. It comes in a granular or powder form and dissolves quickly for efficient cleaning.

  • Form: Granules, powder
  • Container type: Pail
  • Container size: 40 lb
  • Scent: Pine
  • Application: Dissolves quickly and removes tough, embedded soils from concrete floors

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.