Learn More
MSC SC Johnson Professional EZ CARE Heavy Duty Scrub Pad, 13 in Diameter, Red/Gray
Catalog No. 502839998
Encompass
The EZ CARE Heavy Duty Scrub Pad is a durable, dual-sided cleaning pad designed for low-speed scrubbing machines. It effectively removes scuffs, scratches, and black heel marks while reducing burnishing and maintaining the floor's high gloss appearance, cleaning and shining in a single step.
  • Long lasting, dual-sided construction
  • Effectively removes scuffs, scratches, and black heel marks
  • Reduces burnishing and maintains the floor's high gloss appearance
  • Cleans and shines in the same step
  • Red side easily adheres to equipment
  • Gray side for more abrasive cleaning of coated surfaces
  • Made from a proprietary blend of abrasives and resin
  • Designed for use with rotary floor machines (175-300 RPM)
  • Packaged 5 per carton
Catalog No. 50-283-9998 Supplier MSC Supplier No. 95360590
The EZ CARE Heavy Duty Scrub Pad is a durable, dual-sided cleaning pad designed for low-speed scrubbing machines. It effectively removes scuffs, scratches, and black heel marks while reducing burnishing and maintaining the floor's high gloss appearance, cleaning and shining in a single step.
  • Long lasting, dual-sided construction
  • Effectively removes scuffs, scratches, and black heel marks
  • Reduces burnishing and maintains the floor's high gloss appearance
  • Cleans and shines in the same step
  • Red side easily adheres to equipment
  • Gray side for more abrasive cleaning of coated surfaces
  • Made from a proprietary blend of abrasives and resin
  • Designed for use with rotary floor machines (175-300 RPM)
  • Packaged 5 per carton