Learn More
MSC HD WELDING HELMET
Catalog No. 5000781721
Encompass
This welding helmet is designed for protective use, featuring a durable black PPA construction. Polyphthalamide (PPA) is a thermoplastic synthetic resin known for its lightweight properties and resistance to chemicals, heat, and scratches. The helmet incorporates a lift front window style for user convenience.
  • Lift front window style
  • Made of PPA material
  • Lightweight and resistant to chemicals, heat, and scratches
  • Window width: 6.6929 in
  • Window height: 4.0945 in
  • Helmet color: Black
  • Includes rigid neck cover
  • Lens not included
Catalog No. 50-007-81721 Supplier MSC Supplier No. 18477968
This welding helmet is designed for protective use, featuring a durable black PPA construction. Polyphthalamide (PPA) is a thermoplastic synthetic resin known for its lightweight properties and resistance to chemicals, heat, and scratches. The helmet incorporates a lift front window style for user convenience.
  • Lift front window style
  • Made of PPA material
  • Lightweight and resistant to chemicals, heat, and scratches
  • Window width: 6.6929 in
  • Window height: 4.0945 in
  • Helmet color: Black
  • Includes rigid neck cover
  • Lens not included