Learn More
GENIE GROUP HD450 Light Meter, Data Logging, NIST
Catalog No. NC1668745
Encompass_Preferred
The Extech by FLIR light meter measures from 0 to 40,000Fc and 0 to 400kLux with a 0.01Fc/0.1Lux resolution. Features: 5% accuracy, peak mode (10µs), relative mode, min./max., data hold, backlit LCD display with 40-segment bar graph, and a rugged double molded housing. In addition, the datalogger function can automatically store up to 16,000 readings or manually store/recall up to 99 readings.
Catalog No. NC1668745 Supplier GENIE GROUP Supplier No. HD450NIST
The Extech by FLIR light meter measures from 0 to 40,000Fc and 0 to 400kLux with a 0.01Fc/0.1Lux resolution. Features: 5% accuracy, peak mode (10µs), relative mode, min./max., data hold, backlit LCD display with 40-segment bar graph, and a rugged double molded housing. In addition, the datalogger function can automatically store up to 16,000 readings or manually store/recall up to 99 readings.