Learn More
Apexbio Technology LLC HDAC-Act-Fluoro-Assay-Kit
SDP
Catalog No. 5000567473
Encompass_Preferred
HDAC Activity Fluorometric Assay Kit is an essential tool in biomedical research focused on histone deacetylase (HDAC) enzymes which play a critical role in regulating gene expression by removing acetyl groups from histones affecting chromatin structure and function This kit originates from molecular biology laboratories designed to measure HDAC activity through a fluorescence-based mechanism which detects changes in emitted light It is commonly employed for assessing HDAC activity in vitro with activity typically observed in the nanomolar to low micromolar range Researchers use this assay in various applications such as studying epigenetic regulation in cell models evaluating HDAC inhibitors in drug screening and exploring epigenetic alterations in animal studies Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67473 Supplier Apexbio Technology LLC Supplier No. K2031100
Add to Cart
Add to Cart
HDAC Activity Fluorometric Assay Kit is an essential tool in biomedical research focused on histone deacetylase (HDAC) enzymes which play a critical role in regulating gene expression by removing acetyl groups from histones affecting chromatin structure and function This kit originates from molecular biology laboratories designed to measure HDAC activity through a fluorescence-based mechanism which detects changes in emitted light It is commonly employed for assessing HDAC activity in vitro with activity typically observed in the nanomolar to low micromolar range Researchers use this assay in various applications such as studying epigenetic regulation in cell models evaluating HDAC inhibitors in drug screening and exploring epigenetic alterations in animal studies Experimental concentrations typically depend on the specific research design