Learn More
Enzo Life Sciences HDAC assay buffer (20ml)
SDP
Catalog No. 502011099
Encompass_Preferred
Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 137mM sodium chloride, 2.7mM potassium chloride, and 1mM magnesium chlroide. Long Term Storage: -20°C.
Catalog No. 50-201-1099 Supplier Enzo Life Sciences Supplier No. BMLKI1430020
Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 137mM sodium chloride, 2.7mM potassium chloride, and 1mM magnesium chlroide. Long Term Storage: -20°C.