Learn More
Enzo Life Sciences HDAC assay buffer II (20ml)
SDP
Catalog No. 502011119
Encompass_Preferred
50 mM Tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl2, 1 mg/ml BSA. Long Term Storage: -20°C.
Catalog No. 50-201-1119 Supplier Enzo Life Sciences Supplier No. BMLKI4220020
50 mM Tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl2, 1 mg/ml BSA. Long Term Storage: -20°C.