Learn More
Active Motif HDAC Assay Kit (Fluorescent)
SDP
Catalog No. NC1850394
Encompass
HDAC Assay Kits are 96-well plate-based assays to determine the HDAC (Histone Deacetylase) activity in your extracts or purified samples. HDAC Assay Kits are available in both colorimetric and fluorescent formats to suit your needs.
Catalog No. NC1850394 Supplier Active Motif Supplier No. 56200
HDAC Assay Kits are 96-well plate-based assays to determine the HDAC (Histone Deacetylase) activity in your extracts or purified samples. HDAC Assay Kits are available in both colorimetric and fluorescent formats to suit your needs.