Learn More
Cayman Chemical HDAC Fluorometric Activity Assay Kit, 96 Well
SDP
Catalog No. 502886851
Encompass_Preferred
Cayman’s HDAC Fluorometric Activity Assay Kit provides a fast, fluorometric method for measuring HDAC activity without the use of radioactivity, extraction, or chromatography. In the first step, an acetylated lysine substrate is incubated with samples. Deacetylation sensitizes the substrate such that treatment with the HDAC developer in the second step releases a fluorescent product. Trichostatin A is included as a control inhibitor. The fluorophore can be easily analyzed using a fluorescence plate reader with an excitation wavelength of 340-360 nm and an emission wavelength of 440-465 nm. The assay can be used for quantifying Class I and II HDAC activity from various sources.
Catalog No. 50-288-6851 Supplier Cayman Chemical Supplier No. 1001156396 WELL
Cayman’s HDAC Fluorometric Activity Assay Kit provides a fast, fluorometric method for measuring HDAC activity without the use of radioactivity, extraction, or chromatography. In the first step, an acetylated lysine substrate is incubated with samples. Deacetylation sensitizes the substrate such that treatment with the HDAC developer in the second step releases a fluorescent product. Trichostatin A is included as a control inhibitor. The fluorophore can be easily analyzed using a fluorescence plate reader with an excitation wavelength of 340-360 nm and an emission wavelength of 440-465 nm. The assay can be used for quantifying Class I and II HDAC activity from various sources.