Learn More
Medchemexpress LLC HDAC-IN-27 dihydrochloride | 3069831-25-7 | 423.34 g/mol | C20H24Cl2N4O2 | 5 MG
SDP
Catalog No. 5000221278
Encompass_Preferred
HDAC-IN-27 dihydrochloride is a small-molecule class I histone deacetylase (HDAC) inhibitor used in biochemical and cellular research. Supplied as the dihydrochloride salt, it shows activity against HDAC1, HDAC2, and HDAC3 and is applied in studies of apoptosis and oncology models.
  • Class I HDAC-selective inhibitor with demonstrated potency against HDAC1, HDAC2, and HDAC3.
  • Supplied as a dihydrochloride salt for improved stability and solubility.
  • Suitable for biochemical, cellular, and preclinical in vivo studies.
  • Applicable to apoptosis and cancer research, including acute myeloid leukemia models.
  • Available in small lab-scale pack sizes for research use.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-21278 Supplier Medchemexpress LLC Supplier No. HY142690A5MG
HDAC-IN-27 dihydrochloride is a small-molecule class I histone deacetylase (HDAC) inhibitor used in biochemical and cellular research. Supplied as the dihydrochloride salt, it shows activity against HDAC1, HDAC2, and HDAC3 and is applied in studies of apoptosis and oncology models.
  • Class I HDAC-selective inhibitor with demonstrated potency against HDAC1, HDAC2, and HDAC3.
  • Supplied as a dihydrochloride salt for improved stability and solubility.
  • Suitable for biochemical, cellular, and preclinical in vivo studies.
  • Applicable to apoptosis and cancer research, including acute myeloid leukemia models.
  • Available in small lab-scale pack sizes for research use.
  • For research use only; not for human or clinical use.