Learn More
Medchemexpress LLC HDAC-IN-4 | 1252003-13-6 | 50 MG
SDP
Catalog No. 5000341070
Encompass_Preferred
HDAC-IN-4 is a selective inhibitor targeting both HDAC6 and HDAC10, exhibiting pIC50 values of 7.2 and 6.8 in BRET assays, respectively. This compound has demonstrated antitumoral activity.
  • Selective HDAC6 and HDAC10 inhibitor
  • Exhibits antitumoral activity
  • Used in BRET assays
Catalog No. 50-003-41070 Supplier Medchemexpress LLC Supplier No. HY12876350MG
HDAC-IN-4 is a selective inhibitor targeting both HDAC6 and HDAC10, exhibiting pIC50 values of 7.2 and 6.8 in BRET assays, respectively. This compound has demonstrated antitumoral activity.
  • Selective HDAC6 and HDAC10 inhibitor
  • Exhibits antitumoral activity
  • Used in BRET assays