Learn More
Medchemexpress LLC HDAC-IN-4 | 1252003-13-6 | 98.0% | 10 MG
SDP
Catalog No. 5000339323
Encompass_Preferred
HDAC-IN-4 is a selective inhibitor of HDAC6 and HDAC10, exhibiting antitumoral activity. It also inhibits HDAC1, HDAC2, HDAC3, HDAC8, and HDAC10, and is noted for its potency against HDAC6.
  • Selective HDAC6 and HDAC10 inhibitor
  • Exhibits antitumoral activity
  • Inhibits HDAC1, HDAC2, HDAC3, HDAC8, and HDAC10
  • More potent against HDAC6 than Tubastatin A
Catalog No. 50-003-39323 Supplier Medchemexpress LLC Supplier No. HY12876310MG
HDAC-IN-4 is a selective inhibitor of HDAC6 and HDAC10, exhibiting antitumoral activity. It also inhibits HDAC1, HDAC2, HDAC3, HDAC8, and HDAC10, and is noted for its potency against HDAC6.
  • Selective HDAC6 and HDAC10 inhibitor
  • Exhibits antitumoral activity
  • Inhibits HDAC1, HDAC2, HDAC3, HDAC8, and HDAC10
  • More potent against HDAC6 than Tubastatin A