Learn More
Medchemexpress LLC Hdac-in-4 | 1252003-13-6 | 98.0% | 335.40 g·mol-1 | C20H21N3O2 | 5 MG
SDP
Catalog No. 5000262073
Encompass_Preferred
HDAC-IN-4 is a small-molecule inhibitor selective for histone deacetylase isoforms HDAC6 and HDAC10, with reported cellular activity in BRET and FRET assays and demonstrated antitumoral effects in preclinical studies. Supplied as a solid in multiple milligram sizes or as a 10 mM solution in DMSO, it is intended for research use in biochemical and cellular assays.
  • Selective inhibition of HDAC6 and HDAC10.
  • pIC50 values of 7.2 (HDAC6) and 6.8 (HDAC10) in BRET assays.
  • Observed activity in FRET assays against several HDAC isoforms.
  • Available as small solid quantities and as a 10 mM solution in DMSO.
  • High purity (98.0%) suitable for biochemical assays.
  • Molecular weight 335.40 g·mol-1 and formula C20H21N3O2.
Catalog No. 50-002-62073 Supplier Medchemexpress LLC Supplier No. HY1287635MG
HDAC-IN-4 is a small-molecule inhibitor selective for histone deacetylase isoforms HDAC6 and HDAC10, with reported cellular activity in BRET and FRET assays and demonstrated antitumoral effects in preclinical studies. Supplied as a solid in multiple milligram sizes or as a 10 mM solution in DMSO, it is intended for research use in biochemical and cellular assays.
  • Selective inhibition of HDAC6 and HDAC10.
  • pIC50 values of 7.2 (HDAC6) and 6.8 (HDAC10) in BRET assays.
  • Observed activity in FRET assays against several HDAC isoforms.
  • Available as small solid quantities and as a 10 mM solution in DMSO.
  • High purity (98.0%) suitable for biochemical assays.
  • Molecular weight 335.40 g·mol-1 and formula C20H21N3O2.