Learn More
Medchemexpress LLC HDAC-IN-4 | >98.0% | 100 MG
SDP
Catalog No. 5000342929
Encompass_Preferred
HDAC-IN-4 is a selective inhibitor of Histone Deacetylase (HDAC) 6 and HDAC10. It demonstrates potent inhibitory activity against these enzymes, with pIC50 values of 7.2 for HDAC6 and 6.8 for HDAC10 in BRET assays.
  • Selective HDAC6 and HDAC10 inhibitor
  • Antitumoral activity
  • pIC50 of 7.2 for HDAC6 in BRET assay
  • pIC50 of 6.8 for HDAC10 in BRET assay
Catalog No. 50-003-42929 Supplier Medchemexpress LLC Supplier No. HY128763100MG
HDAC-IN-4 is a selective inhibitor of Histone Deacetylase (HDAC) 6 and HDAC10. It demonstrates potent inhibitory activity against these enzymes, with pIC50 values of 7.2 for HDAC6 and 6.8 for HDAC10 in BRET assays.
  • Selective HDAC6 and HDAC10 inhibitor
  • Antitumoral activity
  • pIC50 of 7.2 for HDAC6 in BRET assay
  • pIC50 of 6.8 for HDAC10 in BRET assay