Learn More
TARGETMOL CHEMICALS INC HDAC-IN-40 5MG
SDP
Catalog No. 502579679
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. HDAC-IN-40 is a potent alkoxyamide-based HDAC inhibitor with Ki of 60 nM and 30 nM for HDAC2 and HDAC6 respectively. HDAC-IN-40 had antitumor effectspurity: 99%
Catalog No. 50-257-9679 Supplier TARGETMOL CHEMICALS INC Supplier No. T600825MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. HDAC-IN-40 is a potent alkoxyamide-based HDAC inhibitor with Ki of 60 nM and 30 nM for HDAC2 and HDAC6 respectively. HDAC-IN-40 had antitumor effectspurity: 99%