Learn More
Cell Signaling Technology HDAC1 (10E2) Mouse Monoclonal Antibody, HRP Conjugate, 100 μL
SDP
Catalog No. 502732703
Encompass
This mouse monoclonal antibody is conjugated to horseradish peroxidase (HRP) through its carbohydrate and amine groups. It is specifically designed to detect endogenous levels of total HDAC1 protein and does not cross-react with other HDAC proteins. The antibody is produced by immunizing animals with a synthetic peptide corresponding to the carboxy terminus of the human HDAC1 protein.
  • Validated for western blotting applications.
  • Species reactivity confirmed in human, mouse, rat, and monkey samples.
  • Supplied in a formulation containing 136 mM NaCl, 2.6 mM KCl, 12 mM sodium phosphate (pH 7.4) dibasic, 2 mg/ml BSA, and 50% glycerol.
  • Requires storage at –20°C without aliquoting.
  • Features a concentration of 270 μg/ml.
Catalog No. 50-273-2703 Supplier Cell Signaling Technology Supplier No. 59581S
This mouse monoclonal antibody is conjugated to horseradish peroxidase (HRP) through its carbohydrate and amine groups. It is specifically designed to detect endogenous levels of total HDAC1 protein and does not cross-react with other HDAC proteins. The antibody is produced by immunizing animals with a synthetic peptide corresponding to the carboxy terminus of the human HDAC1 protein.
  • Validated for western blotting applications.
  • Species reactivity confirmed in human, mouse, rat, and monkey samples.
  • Supplied in a formulation containing 136 mM NaCl, 2.6 mM KCl, 12 mM sodium phosphate (pH 7.4) dibasic, 2 mg/ml BSA, and 50% glycerol.
  • Requires storage at –20°C without aliquoting.
  • Features a concentration of 270 μg/ml.