Learn More
Aviva Systems Biology Corp HDAC1 Antibody - C-terminal region (ARP38530_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38530_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HDAC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HDAC1. Alternate names: HD1, RPD3, GON-10, RPD3L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-56690