Learn More
Enzo Life Sciences HDAC1 (human), (recombinant) (His-tag) (50 µg)
SDP
Catalog No. 502012034
Encompass_Preferred
Alternative name: Histone deacetylase 1. Formulation: Liquid. In 50mM TRIS, pH 8.0, 138mM sodium chloride and 10% glycerol. Source: Produced in insect cells. HDAC1 from human cDNA (482 aa). Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2034 Supplier Enzo Life Sciences Supplier No. BMLSE4560050
Alternative name: Histone deacetylase 1. Formulation: Liquid. In 50mM TRIS, pH 8.0, 138mM sodium chloride and 10% glycerol. Source: Produced in insect cells. HDAC1 from human cDNA (482 aa). Produced in a baculovirus expression system. Long Term Storage: -80°C.